Learn More
Invitrogen™ HAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596526
Description
Immunogen sequence: TPFQGQGHGA QLLETVHRYY TEFPTVLDIT AEDPSKSYVK LRDFVLVKLC QDLPCFSREK LMQGFNEDMA IEAQQKFKIN KQHARRVYEI LRLLVTDMSD AEQYRSYRLD IKRRLISPYK KKQRDLAKMR KCLRPEELTN QMNQIEISMQ HEQLEESFQE LVEDYRRVIE RLAQE; Positive Samples: MCF7, SW480, Jurkat, 293T, HeLa, HepG2, U-251MG, Mouse spleen, Rat testis; Cellular Location: Cytoplasm, Nucleus, Nucleus matrix, nucleoplasm.
The protein encoded by this gene is a type B histone acetyltransferase (HAT) that is involved in the rapid acetylation of newly synthesized cytoplasmic histones, which are in turn imported into the nucleus for de novo deposition onto nascent DNA chains. Histone acetylation, particularly of histone H4, plays an important role in replication-dependent chromatin assembly. Specifically, this HAT can acetylate soluble but not nucleosomal histone H4 at lysines 5 and 12, and to a lesser degree, histone H2A at lysine 5. Alternatively spliced transcript variants have been identified for this gene.
Specifications
HAT1 | |
Polyclonal | |
Unconjugated | |
HAT1 | |
2410071B14Rik; AA536933; HAT1; Hat-1; histidine aminotransferase 1; histone acetyltransferase 1; Histone acetyltransferase type B catalytic subunit; histone aminotransferase 1; KAT1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
107435, 296501, 8520 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14929, Q5M939, Q8BY71 | |
HAT1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 245-419 of human KAT1/KAT1/HAT1 (NP_0036331). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.