Learn More
Invitrogen™ HAUS1 Monoclonal Antibody (1E3)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA522937
Catalog No. PIMA522937
Only null left
Add to Cart
Description
Description
Peptide Sequence: RQNMDFLKAK SEEFRFGIKA AEEQLSARGM DASLSHQSLV ALSEKLARLK QQTIPLKKKL ESYLDLMPNP SLAQVKIEEA KRELDSIEAE LTRRVDMMEL.
Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HAUS1
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q96CS2
HAUS1
HAUS1 (NP_612452.1, 179 a.a. ~ 278 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot
1E3
Unconjugated
HAUS1
BC024400; CCDC5; coiled-coil domain containing 5; coiled-coil domain containing 5 (spindle associated); coiled-coil domain-containing protein 5; Enhancer of invasion-cluster; HAUS augmin like complex subunit 1; HAUS augmin-like complex subunit 1; HAUS augmin-like complex, subunit 1; HAUS1; HEIC; HEI-C; HsT1461; spindle associated
Mouse
Affinity chromatography
RUO
115106
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.