Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAUS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117978
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117978 Supplier Invitrogen™ Supplier No. PA5117978
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HAUS1 Polyclonal Antibody for Western Blot, ICC/IF, IP

Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAUS1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene HAUS1
Gene Accession No. Q96CS2
Gene Alias BC024400; CCDC5; coiled-coil domain containing 5; coiled-coil domain containing 5 (spindle associated); coiled-coil domain-containing protein 5; Enhancer of invasion-cluster; HAUS augmin like complex subunit 1; HAUS augmin-like complex subunit 1; HAUS augmin-like complex, subunit 1; HAUS1; HEIC; HEI-C; HsT1461; spindle associated
Gene Symbols HAUS1
Host Species Rabbit
Immunogen E. coli-derived Human HAUS1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115106
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.