Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAUS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534874
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534874 Supplier Invitrogen™ Supplier No. PA534874
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted reactivity: Mouse (84%), Rat (85%), Pig (88%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAUS1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene HAUS1
Gene Accession No. Q96CS2
Gene Alias BC024400; CCDC5; coiled-coil domain containing 5; coiled-coil domain containing 5 (spindle associated); coiled-coil domain-containing protein 5; Enhancer of invasion-cluster; HAUS augmin like complex subunit 1; HAUS augmin-like complex subunit 1; HAUS augmin-like complex, subunit 1; HAUS1; HEIC; HEI-C; HsT1461; spindle associated
Gene Symbols HAUS1
Host Species Rabbit
Immunogen Full length human HAUS1 Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115106
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.