Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAUS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559013 Supplier Invitrogen™ Supplier No. PA559013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IPHLAANLKK MNQALAKMDI LVTETEELAE NILKWRKQQN EVSSCIPKIL AEESYLYKHD IIMPPLPFTS KVHVQTINAK Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 65%.

HAUS2 is 1 of 8 subunits of the 390-kD human augmin complex, or HAUS complex. The augmin complex was first identified in Drosophila, and its name comes from the Latin verb augmentare, meaning to increase. The augmin complex is a microtubule-binding complex involved in microtubule generation within the mitotic spindle and is vital to mitotic spindle assembly (Goshima et al., 2008 [PubMed 18443220]; Uehara et al., 2009 [PubMed 19369198]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAUS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HAUS2
Gene Accession No. Q9NVX0
Gene Alias C15orf25; centrosomal protein 27kDa; centrosomal protein of 27 kDa; Cep27; HAUS augmin like complex subunit 2; HAUS augmin-like complex subunit 2; HAUS augmin-like complex, subunit 2; HAUS2; HsT17025
Gene Symbols HAUS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HAUS2. Recombinant protein control fragment (Product #RP-98069).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55142
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.