Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAUS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561685
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561685 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561685 Supplier Invitrogen™ Supplier No. PA561685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSCGNEFVET LKKIGYPKAD NLNGEDFDWL FEGVEDESFL KWFCGNVNEQ NVLSERELEA FSILQKSGKP ILEGAALDEA LKTCKTSDLK TPRLDDKELE KLEDEVQTLL KLKNLK Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 82%.

Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAUS3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HAUS3
Gene Accession No. Q68CZ6
Gene Alias C4orf15; D4S43h; D5H4S43; D5H4S43E; dgt3; HAUS augmin like complex subunit 3; HAUS augmin-like complex subunit 3; HAUS augmin-like complex, subunit 3; HAUS3; IT1
Gene Symbols HAUS3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HAUS3. Recombinant protein control fragment (Product #RP-98207).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79441
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.