Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HAUS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565275
Description
Immunogen sequence: IHEVVESSNED NFQLLDIQTP SICDNQEILE ERRLEMARLQ LAYICAQHQL IHLKASNSSM KSSIKWAEES LHSLTSKAVD KENLDAKISS LTSEIM Highest antigen sequence identity to the following orthologs - mouse 78%, rat 84%.
Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
Specifications
HAUS3 | |
Polyclonal | |
Unconjugated | |
HAUS3 | |
C4orf15; D4S43h; D5H4S43; D5H4S43E; dgt3; HAUS augmin like complex subunit 3; HAUS augmin-like complex subunit 3; HAUS augmin-like complex, subunit 3; HAUS3; IT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79441 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68CZ6 | |
HAUS3 | |
Recombinant Human HAUS3. Recombinant protein control fragment (Product #RP-105991). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction