Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HAUS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528494
Description
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (83%), Rat (84%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a subunit of the centrosome complex termed the human augmin complex. The encoded protein localizes to the spindle microtubules and may play a role in mitotic spindle assembly and maintenance of centrosome integrity during cell division. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 1.
Specifications
HAUS4 | |
Polyclonal | |
Unconjugated | |
Haus4 | |
9430093H08Rik; AU016300; C14orf94; D14Ertd500e; HAUS augmin like complex subunit 4; HAUS augmin-like complex subunit 4; HAUS augmin-like complex, subunit 4; HAUS4; homologous to Augmin subunits; RGD1305288 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54930 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9H6D7 | |
Haus4 | |
Recombinant protein encompassing a sequence within the center region of human HAUS4. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction