Learn More
Invitrogen™ HAUS7 Monoclonal Antibody (OTI2E9)

Description
HAUS7 Monoclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | HAUS7 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2E9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HAUS7 |
| Gene Accession No. | Q99871 |
| Gene Alias | 1110020L19Rik; 26S proteasome-associated UCH37-interacting protein 1; HAUS augmin like complex subunit 7; HAUS augmin-like complex subunit 7; HAUS augmin-like complex, subunit 7; Haus7; RGD1562991; UCH37-interacting protein 1; UCHL5 interacting protein; UCHL5-interacting protein; UCHL5IP; UIP1; X-linked protein STS1769 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.