Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HAUS7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119172
This item is not returnable.
View return policy
Description
HAUS7 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.
Specifications
HAUS7 | |
Polyclonal | |
Unconjugated | |
HAUS7 | |
1110020L19Rik; 26S proteasome-associated UCH37-interacting protein 1; HAUS augmin like complex subunit 7; HAUS augmin-like complex subunit 7; HAUS augmin-like complex, subunit 7; Haus7; RGD1562991; UCH37-interacting protein 1; UCHL5 interacting protein; UCHL5-interacting protein; UCHL5IP; UIP1; X-linked protein STS1769 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
55559 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q99871 | |
HAUS7 | |
Recombinant protein HAUS Augmin Like Complex Subunit 7. The antigen corresponds to amino acid range 4-319 of the target protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction