Learn More
HAUS7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305557AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.Specifications
HAUS7 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q99871 | |
HAUS7 | |
Between 318 and 368 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
HAUS7 | |
HAUS augmin-like complex, subunit 7, HAUS7, UCHL5IP, UIP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55559 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.