Learn More
Haws™ AXION Advantage™ Upgrade Kit
SDP
Catalog No. p-7138091 Shop All Haws Corporation Products
Click to view available options
Includes:
Eye and Face Wash Head
Eye, Face Wash Head and Showerhead
Lab Eye and Face Wash Head
Material:
Plastic
Stainless Steel
Haws™ AXION Advantage™ Upgrade Kit provides Zero Vertical Velocity™ stream engineering with even laminar flow for enhanced comfort, stability and effectiveness.
  • Designed to replace outdated, under-performing or in some cases, non-conforming installed emergency eyewashes with the medically superior AXION™ MSR performance.
  • This revolutionary inverted flow design is the only product on the market that provides a Medically Superior Response consistent with all EMT, emergency room and doctors' office protocols by sweeping contaminants away from the vulnerable nasal cavity.
  • Traditional eyewashes irrigate from the outside edge of the eye back toward the nose, where contaminants can then be flushed into the nasal cavity through the lacrimal punctum, nature's ocular cavity drain.
  • Eyewash streams provide Zero Vertical Velocity™ stream engineering with even laminar flow for enhanced comfort, stability and effectiveness.
  • System is designed to replace the eyewash head of some standard laboratory and hose-mounted eyewash stations manufactured by Acorn™, Bradley™, Encon™, Guardian and Speakman™.
The AXION Advantage system is ANSI compliant when installed correctly based on manufacturer guidelines and when tested against the ANSI Z358.1 checklist included with the product. The materials provided offer complete ANSI compliance tools and instructions. At the current time, ANSI does not regulate product brands, models, or the interchange of product components.
Warranty and Services
1 year from ship date (Limited)
Specifications
Color Green
Includes Eye and Face Wash Head
Material Plastic
Product Type Eyewash Upgrade Kit
Weight (English) 3 lb.
Weight (Metric) 1.36078 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.