Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Freeze Valve
SDP

Catalog No. p-7138094 Shop All Haws Corporation Products
Click to view available options
For Use With (Equipment):
Emergency Showers
Emergeny Shower and Eyewash

Haws™ Freeze Valve is a retro-fit freeze protection valve kit

Model SP158.8, retro-fit freeze valve kit to replace the stainless steel freeze valve on Haws model 8300.158. This kit is supplied with the necessary hardware and instructions.

Warranty and Services

1 year from ship date (Limited)

Specifications

For Use With (Equipment) Emergeny Shower and Eyewash
Material Brass
Product Type Freeze Valve
Weight (English) 3 lb.
Weight (Metric) 1.36 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.