Learn More
Invitrogen™ HAX1 Polyclonal Antibody

Description
Recommended positive controls: Molt-4, Raji, 293T, A431 , HeLa, HepG2. Predicted reactivity: Mouse (80%), Rhesus Monkey (97%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | HAX1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.87 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | Hax1 |
| Gene Accession No. | O00165, O35387, Q7TSE9 |
| Gene Alias | FLJ17042; FLJ18492; FLJ93803; Hax1; HAX-1; HCLS1 (and PKD2) associated protein; HCLS1 associated protein X-1; HCLS1 associated X-1; HCLS1-associated protein X-1; HCLSBP1; HS1 binding protein; HS1-associated protein X-1; HS1-associating protein X-1; HS1-binding protein 1; HS1BP1; HSP1BP-1; mHAX-1s; OTTHUMP00000034190; RP11-137P24.1; SCN3; SIG-111; Silg111 |
| Gene Symbols | Hax1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.