Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAX1 Recombinant Rabbit Monoclonal Antibody (HL2170)
GREENER_CHOICE

Catalog No. FEMA550416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550416 Supplier Invitrogen™ Supplier No. MA550416
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

HAX1 Recombinant Monoclonal Antibody for IHC (P), Western Blot

The protein encoded by this gene is known to associate with hematopoietic cell-specific Lyn substrate 1, a substrate of Src family tyrosine kinases. It also interacts with the product of the polycystic kidney disease 2 gene, mutations in which are associated with autosomal-dominant polycystic kidney disease, and with the F-actin-binding protein, cortactin. It was earlier thought that this gene product is mainly localized in the mitochondria, however, recent studies indicate it to be localized in the cell body. Mutations in this gene result in autosomal recessive severe congenital neutropenia, also known as Kostmann disease. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2170
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Hax1
Gene Accession No. O00165, O35387
Gene Alias FLJ17042; FLJ18492; FLJ93803; Hax1; HAX-1; HCLS1 (and PKD2) associated protein; HCLS1 associated protein X-1; HCLS1 associated X-1; HCLS1-associated protein X-1; HCLSBP1; HS1 binding protein; HS1-associated protein X-1; HS1-associating protein X-1; HS1-binding protein 1; HS1BP1; HSP1BP-1; mHAX-1s; OTTHUMP00000034190; RP11-137P24.1; SCN3; SIG-111; Silg111
Gene Symbols Hax1
Host Species Rabbit
Immunogen Recombinant fragmemt of human HAX1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10456, 23897
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.