Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAX1Rabbit anti-Chicken, Human, Mouse, Non-human primate, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501992255
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2255 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2255 Supplier Rockland Immunochemicals Supplier No. 600401J67
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-HAX1 was affinity purified from monospecific antiserum by immunoaffinity chromatography. A BLAST analysis was used to suggest cross-reactivity with human, rat, and mouse based on 100% sequence homology. Cross-reactivity with HAX1 from other sources has not been determined.

The protein encoded by this gene is known to associate with hematopoietic cell-specific Lyn substrate 1, a substrate of Src family tyrosine kinases. It also interacts with the product of the polycystic kidney disease 2 gene, mutations in which are associated with autosomal-dominant polycystic kidney disease, and with the F-actin-binding protein, cortactin. It was earlier thought that this gene product is mainly localized in the mitochondria, however, recent studies indicate it to be localized in the cell body. Mutations in this gene result in autosomal recessive severe congenital neutropenia, also known as Kostmann disease. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen HAX1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.78 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl, 30% glycerol and no preservative; pH 7.2
Gene Hax1
Gene Accession No. O00165, O35387, Q7TSE9
Gene Alias FLJ17042; FLJ18492; FLJ93803; HAX1; HAX-1; HCLS1 (and PKD2) associated protein; HCLS1 associated protein X-1; HCLS1 associated X-1; HCLS1-associated protein X-1; HCLSBP1; HS1 binding protein; HS1-associated protein X-1; HS1-associating protein X-1; HS1-binding protein 1; HS1BP1; HSP1BP-1; mHAX-1s; OTTHUMP00000034190; RP11-137P24.1; SCN3; SIG-111; Silg111
Gene Symbols Hax1
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10456, 107055138, 23897, 291202
Target Species Chicken, Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.