Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hayem Diluting Fluid, for Erythrocyte (Red Cell) Count, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
| Physical Form | Liquid |
| Chemical Name or Material | Hayem Diluting Fluid |
| CAS | 7732-18-5, 7757-82-6, 7647-14-5, 7487-94-7 |
| CAS Min % | 95.0, 2.37, 0.47, 0.24 |
| CAS Max % | 99.0, 2.47, 0.49, 0.25 |
| Packaging | Amber Glass Bottle |
| Color | Colorless |
| Quantity | 1 L |
| Grade | Laboratory |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction