Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Hazardous Material Label: ISOPROPYL ACETATE
Shop All Brady Corporation Products
Click to view available options
Display Text:
ISOPROPYL ACETATE
Description
- Provides workers with chemical information of hazardous materials
- Made from B-928 vinyl material
- Label Size: W x H: 4.5 x 3.75 in.
Specifications
Specifications
Adhesive Type | Acrylic, Permanent, Cold-temperature |
Color | Black, Yellow |
Material | Vinyl (B-928) |
Product Type | Hazardous Materials Label |
Display Text | ISOPROPYL ACETATE |
Quantity | 25/Pk. |
Shape | Rectangle |
Size (English) | 4.5 x 3.75 in. (W x H) |
Thickness (English) | 0.0049 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction