Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Provides workers with chemical information of hazardous materials
- Made from B-928 vinyl material
- Label Size: W x H: 4.5 x 3.75 in.
Specifications
Specifications
Adhesive Type | Acrylic, Permanent, Cold-temperature |
Color | Black, Yellow |
Material | Vinyl (B-928) |
Product Type | Hazardous Materials Label |
Display Text | PHOSPHORYL CHLORIDE |
Quantity | 25/Pk. |
Shape | Rectangle |
Size (English) | 4.5 x 3.75 in. (W x H) |
Thickness (English) | 0.0049 in. |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction