Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Hazardous Material Shipping Labels - Adhesive Vinyl
Shop All Brady Corporation Products
Click to view available options
Display Text:
ORGANIC PEROXIDE 5.2
OXYGEN 2
RADIOACTIVE II CONTENTS: ACTIVITY: TRANSPORT INDEX
Description
- Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
- Up to 3 year average outdoor durability
Specifications
Specifications
Autoclavable | Not Autoclavable |
Certifications/Compliance | ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145 |
Description | Label |
Adhesive Type | Self-adhesive |
Color | Black/Red/Yellow on White |
Format | Self-adhesive |
Material | Vinyl |
Printable Sides | Single-side Printable |
Product Type | Label |
Display Text | RADIOACTIVE II CONTENTS: ACTIVITY: TRANSPORT INDEX |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction