Learn More
Brady™ Hazardous Waste Label: HAZARDOUS WASTE ACCUMULATION START DATE CONTENTS...
Shop All Brady Corporation ProductsDescription
- To identify containers, cylinders and drums holding hazardous waste
- Made of B-7569 vinyl
- Available in size (W x H): 15.2 x 10.16cm (6 x 4 in.)
Specifications
Specifications
| Adhesive Type | Acrylic, Permanent |
| Color | Black, Red, White |
| Material | Vinyl (B-7569) |
| Product Type | Hazardous Waste Label |
| Display Text | HAZARDOUS WASTE ACCUMULATION START DATE CONTENTS...etc |
| Quantity | 50/Pk. |
| Shape | Square |
| Size (English) | 6 x 4 in. (W x H) |
| Thickness (English) | 0.004 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.