Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Waste Label: NON-REGULATED LABEL

Catalog No. p-7070649 Shop All Brady Corporation Products
Click to view available options
Quantity:
100/Pk.
50/Pk.

Identify hazardous waste materials for transportation off-site. Brady™ Hazardous Waste Labels are made of vinyl and comply with all current federal regulations regarding identification of hazardous waste material transportation.

  • To identify containers, cylinders and drums holding hazardous waste
  • Made of B-7569 vinyl
  • Available in size (W x H): 15.2 x 15.2cm (6 x 6 in.)

Specifications

Adhesive Type Acrylic, Permanent
Color White, Blue
Material Vinyl (B-7569)
Product Type Hazardous Waste Label
Display Text NON-REGULATED WASTE
Quantity 100/Pk.
Shape Square
Size (English) 6 x 6 in.
Thickness (English) 0.004 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.