Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ HAZWIK™ High Visibility Absorbent Safety Pads

Catalog No. 01-670-139
Click to view available options
Packaging Quantity:
100 Pads

HAZWIK™ High Visibility Absorbents combine effective absorbency and hazardous-materials messaging in one convenient solution.

  • Surfactant-treated polypropylene picks up a wide range of aggressive chemicals, including hydrofluoric acid
  • Bright color and printed message alert workers to the presence of hazardous materials and waste
  • Green coverstock identifies product for proper separation and disposal, and reduces likelihood of linting or tearing
  • Heavy weight: specifically designed for continuous leaks and larger spills so you use less material
  • Perforations reduce waste by allowing you to use only what you need

Specifications

Color Green
Design Dimpled, Low-linting
Material Polypropylene
Type Absorbent Pad
Packaging Quantity 100 Pads
Length (English) 19 in.
Width (English) 15 in.
Weight (English) 16 lb.
Weight (Metric) 7.2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.