Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HB-EGF Antibody (MM0324-3R44) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50

Specifications

Antigen HB-EGF
Clone MM0324-3R44
Concentration LYOPH
Applications Western Blot, Blocking Assay, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019186
SDP
View Documents
Novus Biologicals
NBP2123520.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP212352
SDP
View Documents
Novus Biologicals
NBP212352
0.1 mg N/A N/A N/A
Description

Description

HB-EGF Monoclonal specifically detects HB-EGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.
Specifications

Specifications

HB-EGF
LYOPH
Monoclonal
Purified
RUO
Q53H93
1839
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
MM0324-3R44
Western Blot, Blocking Assay, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor
HBEGF
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.