Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HB-EGF Rabbit anti-Human, Rat, Clone: S08-9C2, Novus Biologicals™

Rabbit Monoclonal Antibody
$286.00 - $573.00
Specifications
Antigen | HB-EGF |
---|---|
Clone | S08-9C2 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunofluorescence 5 μg/mL, Immunocytochemistry 5 μg/mL |
Applications | Western Blot, ELISA, Immunofluorescence, Immunocytochemistry |
Classification | Monoclonal |
Description
HB-EGF Monoclonal antibody specifically detects HB-EGF in Human, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, ImmunocytochemistrySpecifications
HB-EGF | |
Western Blot, ELISA 1:100-1:2000, Immunofluorescence 5 μg/mL, Immunocytochemistry 5 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor | |
A synthetic peptide of human HB-EGF (Uniprot#: Q99075) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S08-9C2 | |
Western Blot, ELISA, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
1839 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title