Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HB9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540456 Supplier Invitrogen™ Supplier No. PA540456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AAASGTGGGG GGGGASGGTS GSCSPASSEP PAAPADRLRA ESPSPPRLLA Sequence homology: Dog: 86%; Human: 100%; Mouse: 79%; Pig: 93%; Rat: 79%.

Homeobox protein HB9 is expressed in lymphoid and pancreatic tissues. It is reported to be Putative transcription factor involved in pancreas development and function. Defects in this gene causes Currarino syndrome. The triad of a presacral tumor, sacral agenesis and anorectal malformation constitutes the Currarino syndrome which is caused by dorsal-ventral patterning defects during embryonic development. It is also reported that HB9 is strongly expressed in epidermal basal cells of the outer scale face in tarsometatarsal scale skin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HB9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MNX1
Gene Accession No. P50219
Gene Alias HB9; Hlxb9; homeo box HB9; homeobox gene HB9; homeobox HB9; homeobox protein HB9; homeodomain protein Hb9; HOX SCRA1; HOXHB9; LOW QUALITY PROTEIN: motor neuron and pancreas homeobox protein 1; MNR2; MNX1; motor neuron and pancreas homeobox 1; motor neuron and pancreas homeobox protein 1; SCRA1; zgc:112174
Gene Symbols MNX1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MNX1 (aa 36-85).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3110
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.