Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HB9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568569
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 92%; Zebrafish: 92%.
Homeobox protein HB9 is expressed in lymphoid and pancreatic tissues. It is reported to be Putative transcription factor involved in pancreas development and function. Defects in this gene causes Currarino syndrome. The triad of a presacral tumor, sacral agenesis and anorectal malformation constitutes the Currarino syndrome which is caused by dorsal-ventral patterning defects during embryonic development. It is also reported that HB9 is strongly expressed in epidermal basal cells of the outer scale face in tarsometatarsal scale skin.
Specifications
HB9 | |
Polyclonal | |
Unconjugated | |
MNX1 | |
HB9; Hlxb9; homeo box HB9; homeobox gene HB9; homeobox HB9; homeobox protein HB9; homeodomain protein Hb9; HOX SCRA1; HOXHB9; LOW QUALITY PROTEIN: motor neuron and pancreas homeobox protein 1; MNR2; MNX1; motor neuron and pancreas homeobox 1; motor neuron and pancreas homeobox protein 1; SCRA1; zgc:112174 | |
Rabbit | |
Affinity chromatography | |
RUO | |
15285 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9QZW9 | |
MNX1 | |
synthetic peptide directed towards the middle region of mouse HB9. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction