Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HBAE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572208
Description
HBAE1 Polyclonal Antibody for Western Blot

Specifications
HBAE1 | |
Polyclonal | |
Unconjugated | |
hbae1 | |
ae1; alphae1globin; dZ118J2.6; embryonic alpha globin e1; etID40432.3; globin alphae1; HbA; hba2; HBA2r; HBAA1; hbae1; hbae1.1; hemoglobin alpha embryonic-1; hemoglobin alpha-2 chain; hemoglobin, alpha embryonic 1; hemoglobin, alpha embryonic 1.1; si:by119c3.1; si:by119c3.3; SI:dZ118J2.10; SI:dZ118J2.8; si:xx-119c3.3; wu:fa12a11; wu:fb74f06; zgc:86936 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
30597 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.49 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
hbae1 | |
KLH conjugated synthetic peptide between 46-80 amino acids from the Central region of DANRE hbae1. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction