Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HBB Polyclonal Antibody

Catalog No. PIPA5121924
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121924 20 μL
PIPA5119182 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121924 Supplier Invitrogen™ Supplier No. PA5121924
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HBB Polyclonal Antibody for Western Blot, IHC (P)

The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5'-epsilon-gamma-G-gamma-A-delta-beta-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene HBB
Gene Accession No. 0
Gene Alias beta globin chain; beta-1-globin; Beta-globin; CD113t-C; Hbb; hemoglobin beta; Hemoglobin beta chain; hemoglobin beta chain complex; hemoglobin beta chain, major-form; Hemoglobin beta-1 chain; hemoglobin subunit beta; Hemoglobin subunit beta-1; hemoglobin, beta; III beta-1 globin; III beta-2 globin; LVV-hemorphin-7; Spinorphin
Gene Symbols HBB
Host Species Rabbit
Immunogen Recombinant protein Hemoglobin Beta.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15127
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.