Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBcAg Mouse anti-Human, Clone: 1H8, Bioss
SDP

Catalog No. 501979892
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9892 Supplier Bioss Supplier No. BSM2000M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatitis B Virus Core Antigen (HBcAg) is part of the infectious virion containing an inner 'core particle' enclosing the viral genome. The icosahedral core particle contains 180 or 240 copies of the core protein. HBcAg is one of the three major clinical antigens of hepatitis B virus but disappears early in the course of infection.The hepatitis B virus core antigen (HBcAg) is a highly immunogenic subviral particle and functions as both a T-cell-dependent and a T-cell-independent antigen. Therefore, HBcAg may be a promising candidate target for therapeutic vaccine control of chronic HBV infection.

Specifications

Antigen HBcAg
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1H8
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Mouse
Immunogen Recombinant Hepatitis B Core Antigen (rHBcAg).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.