Learn More
HBE1 Mouse anti-Human, Clone: 2C11G6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | HBE1 |
---|---|
Clone | 2C11G6 |
Concentration | 1.04 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6585
![]() ![]() |
Proteintech Group Inc
661511IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody specifically recognizes the HBE1 and doesn't cross-react with other globin chains.
The epsilon globin gene (HBE) is normally expressed in the embryonic yolk sac: two epsilon chains together with two zeta chains (an alpha-like globin) constitute the embryonic hemoglobin Hb Gower I; two epsilon chains together with two alpha chains form the embryonic Hb Gower II. Both of these embryonic hemoglobins are normally supplanted by fetal, and later, adult hemoglobin. The five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5′-epsilon - G-gamma - A-gamma - delta - beta-3′Specifications
HBE1 | |
1.04 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Epsilon globin, HBE, HBE1, Hemoglobin epsilon, Hemoglobin epsilon chain, Hemoglobin subunit epsilon, hemoglobin, epsilon 1 | |
HBE1 | |
Primary | |
-20°C | |
HBE1 |
2C11G6 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P02100 | |
3046 | |
IgG1 | |
Protein G | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.