Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBE1 Mouse anti-Human, FITC, Clone: 2C11G6, Proteintech
SDP

Catalog No. 501737233 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7233 Supplier Proteintech Group Inc Supplier No. FITC66151100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody specifically recognizes the HBE1 and doesn't cross-react with other globin chains. This antibody is conjugated with FITC.

The epsilon globin gene (HBE) is normally expressed in the embryonic yolk sac: two epsilon chains together with two zeta chains (an alpha-like globin) constitute the embryonic hemoglobin Hb Gower I; two epsilon chains together with two alpha chains form the embryonic Hb Gower II. Both of these embryonic hemoglobins are normally supplanted by fetal, and later, adult hemoglobin. The five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5′-epsilon - G-gamma - A-gamma - delta - beta-3′

Specifications

Antigen HBE1
Applications ELISA
Classification Monoclonal
Clone 2C11G6
Concentration 0.33 mg/mL
Conjugate FITC
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.3
Gene HBE1
Gene Accession No. P02100
Gene Alias HBE1, Hemoglobin epsilon, Hemoglobin epsilon chain
Gene Symbols HBE1
Host Species Mouse
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3046
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.