Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HBE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570002 Supplier Invitrogen™ Supplier No. PA570002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Goat: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 86%; Rat: 93%; Sheep: 86%.

The epsilon globin gene (HBE) is normally expressed in the embryonic yolk sac: two epsilon chains together with two zeta chains (an alpha-like globin) constitute the embryonic hemoglobin Hb Gower I; two epsilon chains together with two alpha chains form the embryonic Hb Gower II. Both of these embryonic hemoglobins are normally supplanted by fetal, and later, adult hemoglobin. The five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5'-epsilon - G-gamma - A-gamma - delta - beta-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBE1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HBE1
Gene Accession No. 0
Gene Alias A-gamma globin; Beta-4 globin; epsilon 1 globin; epsilon globin; epsilon I globin; Epsilon-1-globin; Epsilon-globin; GLNB1; globin B1; HBE; HBE1; HBG1; HBG2; Hemoglobin epsilon chain; Hemoglobin epsilon-1 chain; Hemoglobin epsilon-I chain; hemoglobin subunit epsilon; hemoglobin subunit epsilon 1; Hemoglobin subunit epsilon-1; hemoglobin subunit epsilon-1-like; hemoglobin, epsilon 1; hemoglobin, gamma A; hemoglobin, gamma G; LOC102174495
Gene Symbols HBE1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of rat HBE1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293267
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.