Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HBE1 Recombinant Rabbit Monoclonal Antibody (HL2457)
GREENER_CHOICE

Catalog No. FEMA550538
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550538 Supplier Invitrogen™ Supplier No. MA550538
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

HBE1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

The epsilon globin gene (HBE) is normally expressed in the embryonic yolk sac: two epsilon chains together with two zeta chains (an alpha-like globin) constitute the embryonic hemoglobin Hb Gower I; two epsilon chains together with two alpha chains form the embryonic Hb Gower II. Both of these embryonic hemoglobins are normally supplanted by fetal, and later, adult hemoglobin. The five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5'-epsilon - G-gamma - A-gamma - delta - beta-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBE1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2457
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene HBE1
Gene Accession No. P02100
Gene Alias A-gamma globin; Beta-4 globin; epsilon 1 globin; epsilon globin; epsilon I globin; Epsilon-1-globin; Epsilon-globin; GLNB1; globin B1; HBE; HBE1; HBG1; HBG2; Hemoglobin epsilon chain; Hemoglobin epsilon-1 chain; Hemoglobin epsilon-I chain; hemoglobin subunit epsilon; hemoglobin subunit epsilon 1; Hemoglobin subunit epsilon-1; hemoglobin subunit epsilon-1-like; hemoglobin, epsilon 1; hemoglobin, gamma A; hemoglobin, gamma G; LOC102174495
Gene Symbols HBE1
Host Species Rabbit
Immunogen Synthetic peptide of human Hemoglobin epsilon.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110337495, 293267, 3046
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.