Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBEBP2/GLYCTK Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982873
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2873 Supplier Bioss Supplier No. BS13448R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described.

Specifications

Antigen HBEBP2/GLYCTK
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GLYCTK
Gene Accession No. Q8IVS8
Gene Alias 6230410P16Rik; C430006L12; Glycerate kinase; GLYCTK; HBeAg binding protein 4; HBeAg-binding protein 2; HBeAg-binding protein 4; HBeAgBP4A; HBEBP2; HBEBP4; LP5910
Gene Symbols GLYCTK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HBEBP2/GLYCTK.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132158
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.