Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBG1/2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216806
Description
HBG1/2 Polyclonal specifically detects HBG1/2 in Human samples. It is validated for Western Blot.Specifications
Hemoglobin gamma-2 chain | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
hemoglobin subunit gamma-2, hemoglobin, gamma G, TNCY | |
Rabbit | |
16 kDa | |
0.1 mL | |
Cardiovascular Biology | |
3048 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.71mg/mL | |
Western Blot 1:500-1:3000 | |
P69892 | |
HBG2 | |
Full length human Hemoglobin gamma G Recombinant protein. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction