Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBG1/2 Antibody (SR1114), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | HBG1/2 |
---|---|
Clone | SR1114 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
HBG1/2 Monoclonal antibody specifically detects HBG1/2 in Human samples. It is validated for Western BlotSpecifications
HBG1/2 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
abnormal hemoglobin, A-gamma globin, gamma A hemoglobin, gamma globin, Gamma-1-globin, gamma-2-globin, gamma-globin chain, G-gamma globin Paulinia, Hb F Agamma, hb F Ggamma, HBGA, HBGR, HBG-T1, HBG-T2, Hemoglobin gamma-1 chain, hemoglobin gamma-2 chain, Hemoglobin gamma-A chain, hemoglobin gamma-G chain, hemoglobin subunit gamma-1, hemoglobin subunit gamma-2, hemoglobin, gamma A, hemoglobin, gamma G, hemoglobin, gamma, regulator of, HSGGL1, methemoglobin, PRO2979, TNCY | |
A synthesized peptide derived from human HBG1/2 (Uniprot #: P69891, P69892 ) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1114 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3047 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title