Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBG1 Mouse anti-Human, Clone:M2, Abnova™

Catalog No. 89333051 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-051 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-051 Supplier Abnova Corporation Supplier No. H00003047M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant HBG1.

The gamma globin genes (HBG1 and HBG2) are normally expressed in the fetal liver, spleen and bone marrow. Two gamma chains together with two alpha chains constitute fetal hemoglobin (HbF) which is normally replaced by adult hemoglobin (HbA) at birth. In some beta-thalassemias and related conditions, gamma chain production continues into adulthood. The two types of gamma chains differ at residue 136 where glycine is found in the G-gamma product (HBG2) and alanine is found in the A-gamma product (HBG1). The former is predominant at birth. The order of the genes in the beta-globin cluster is: 5′-epsilon -- gamma-G -- gamma-A -- delta -- beta--3′. [provided by RefSeq]

Specifications

Antigen HBG1
Applications ELISA, Western Blot
Classification Monoclonal
Clone M2
Conjugate Unconjugated
Description hemoglobin, gamma A
Formulation PBS with no preservative; pH 7.4
Gene HBG1
Gene Accession No. BC010913
Gene Alias HBGA, HBGR, HSGGL1, PRO2979
Gene Symbols HBG1
Host Species Mouse
Immunogen HBG1 (AAH10913, 1 a.a. ∽ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3047
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.