Learn More
Invitrogen™ HBG2 Polyclonal Antibody

Description
HBG2 Polyclonal Antibody for Flow, IHC (P), Western Blot
Specifications
Specifications
| Antigen | HBG2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | HBG2 |
| Gene Accession No. | P69892 |
| Gene Alias | abnormal hemoglobin; basic fibroblast growth factor; bFGF; Fgf2; Fgf-2; Fgfb; fibroblast growth factor 2; gamma-2-globin; G-gamma globin Paulinia; Hb F Ggamma; HBG2; HBGF-2; HBG-T1; hemoglobin gamma-2 chain; Hemoglobin gamma-G chain; hemoglobin subunit gamma 2; hemoglobin subunit gamma-2; hemoglobin, gamma G; heparin-binding growth factor 2; methemoglobin; TNCY |
| Gene Symbols | HBG2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.