Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HBG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597822
Description
HBG2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The gamma globin genes (HBG1 and HBG2) are normally expressed in the fetal liver, spleen and bone marrow. Two gamma chains together with two alpha chains constitute fetal hemoglobin (HbF) which is normally replaced by adult hemoglobin (HbA) at birth. In some beta-thalassemias and related conditions, gamma chain production continues into adulthood. The two types of gamma chains differ at residue 136 where glycine is found in the G-gamma product (HBG2) and alanine is found in the A-gamma product (HBG1). The former is predominant at birth. The order of the genes in the beta-globin cluster is: 5'- epsilon -- gamma-G -- gamma-A -- delta -- beta--3'.
Specifications
HBG2 | |
Polyclonal | |
Unconjugated | |
HBG2 | |
abnormal hemoglobin; basic fibroblast growth factor; bFGF; Fgf2; Fgf-2; Fgfb; fibroblast growth factor 2; gamma-2-globin; G-gamma globin Paulinia; Hb F Ggamma; HBG2; HBGF-2; HBG-T1; hemoglobin gamma-2 chain; Hemoglobin gamma-G chain; hemoglobin subunit gamma 2; hemoglobin subunit gamma-2; hemoglobin, gamma G; heparin-binding growth factor 2; methemoglobin; TNCY | |
Rabbit | |
Protein G | |
RUO | |
3048 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P69892 | |
HBG2 | |
Recombinant Human Hemoglobin subunit gamma-2 protein (2-147AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction