Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBLD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318410
This item is not returnable.
View return policy
Description
HBLD1 Polyclonal antibody specifically detects HBLD1 in Human samples. It is validated for Western BlotSpecifications
HBLD1 | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
122961 | |
Store at 4°C. Do not freeze. | |
IgG |
Western Blot | |
1 mg/ml | |
Western Blot 1:2000 - 1:10000 | |
c14_5557, HBLD1, HesB like domain containing 1, HESB-like domain-containing protein 1, iron-sulfur cluster assembly 2 homolog (S. cerevisiae), iron-sulfur cluster assembly 2 homolog, mitochondrial, ISA2 | |
Recognizes region between residues Between between 1 and 50.. | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction