Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBO Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501562075
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2075 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2075 Supplier Bethyl Laboratories, Inc Supplier No. A302225AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat

Component of the HBO1 complex which has a histone H4-specific acetyltransferase activity, a reduced activity toward histone H3 and is responsible for the bulk of histone H4 acetylation in vivo. Through chromatin acetylation it may regulate DNA replication and act as a coactivator of TP53-dependent transcription. Specifically represses AR-mediated transcription. {ECO:0000269; PubMed:10438470, ECO:0000269; PubMed:10930412, ECO:0000269; PubMed:11278932, ECO:0000269; PubMed:16387653}.

Specifications

Antigen HBO
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene KAT7
Gene Accession No. O95251, Q5SVQ0
Gene Alias Histone acetyltransferase MYST2; EC 2.3.1.48; MYST protein 2; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 2; Histone acetyltransferase binding to ORC1; MYST2; HBO1; HBOa
Gene Symbols KAT7
Host Species Rabbit
Immunogen Between 561 and 611
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11143, 217127
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.