Learn More
HBO Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00632T

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda and Orangutan
Component of the HBO1 complex which has a histone H4-specific acetyltransferase activity, a reduced activity toward histone H3 and is responsible for the bulk of histone H4 acetylation in vivo. Through chromatin acetylation it may regulate DNA replication and act as a coactivator of TP53-dependent transcription. Specifically represses AR-mediated transcription. {ECO:0000269; PubMed:10438470, ECO:0000269; PubMed:10930412, ECO:0000269; PubMed:11278932, ECO:0000269; PubMed:16387653}.Specifications
HBO | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O95251 | |
KAT7 | |
Between 125 and 175 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
KAT7 | |
Histone acetyltransferase MYST2; EC 2.3.1.48; MYST protein 2; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 2; Histone acetyltransferase binding to ORC1; MYST2; HBO1; HBOa | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11143 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.