Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBP1 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306527V
This item is not returnable.
View return policy
Description
HBP1 Polyclonal antibody specifically detects HBP1 in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
HBP1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunofluorescence | |
DyLight 405 | |
FLJ16340, High mobility group box transcription factor 1, HMG box transcription factor 1, HMG box-containing protein 1, HMG-box containing protein 1, HMG-box transcription factor 1, USMCC2 | |
Produced in rabbits immunized with purified, recombinant Human HBP1 (O60381-1; Pro208-Phe345). | |
0.1 mL | |
Cardiovascular Biology, Stem Cells | |
26959 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction