Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBQ1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 199971AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-2646


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The antibody is specific to HBQ1.

Theta-globin mRNA is found in human fetal erythroid tissue but not in adult erythroid or other nonerythroid tissue. The theta-1 gene may be expressed very early in embryonic life, perhaps sometime before 5 weeks. Theta-1 is a member of the human alpha-globin gene cluster that involves five functional genes and two pseudogenes. The order of genes is: 5′ - zeta - pseudozeta - mu - pseudoalpha-2 -pseudoalpha-1 - alpha-2 - alpha-1 - theta-1 - 3′. Research supports a transcriptionally active role for the gene and a functional role for the peptide in specific cells, possibly those of early erythroid tissue.
Specifications

Specifications

HBQ1
Polyclonal
Unconjugated
HBQ1
HBQ1, Hemoglobin subunit theta 1, Hemoglobin theta 1 chain, hemoglobin, theta 1, Theta 1 globin
Rabbit
Antigen Affinity Chromatography
RUO
3049
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P09105
HBQ1
Peptide
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.