Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBS1L Antibody (OTI1C1), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271628AF488
Description
HBS1L Monoclonal specifically detects HBS1L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| HBS1L | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HBS1L | |
| Recombinant protein fragment corresponding to amino acids 1-260 of human HBS1L (NP_006611) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1C1 | |
| Western Blot, Immunohistochemistry | |
| DKFZp434g247, ERF3-similar protein, ERFSDKFZp686L13262, HBS1 (S. cerevisiae)-like, HBS1KIAA1038EF-1a, HBS1-like (S. cerevisiae), HBS1-like protein, Hsp70 subfamily B suppressor 1-like protein, HSPC276 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 10767.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction