Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HBS1L Monoclonal Antibody (OTI2A3)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526469

Catalog No. PIMA526469


Only null left
Add to Cart

Description

Description

HBS1L Monoclonal Antibody for Western Blot

This gene encodes a member of the GTP-binding elongation factor family. It is expressed in multiple tissues with the highest expression in heart and skeletal muscle. The intergenic region of this gene and the MYB gene has been identified to be a quantitative trait locus (QTL) controlling fetal hemoglobin level, and this region influnces erythrocyte, platelet, and monocyte counts as well as erythrocyte volume and hemoglobin content. DNA polymorphisms at this region associate with fetal hemoglobin levels and pain crises in sickle cell disease. A single nucleotide polymorphism in exon 1 of this gene is significantly associated with severity in beta-thalassemia/Hemoglobin E. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HBS1L
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y450
HBS1L
Human recombinant protein fragment corresponding to amino acids 1-242 of HBS1L produced in E.coli.
100 μL
Primary
Human, Monkey
Antibody
IgG1
Western Blot
OTI2A3
Unconjugated
HBS1L
2810035F15Rik; AI326327; EF-1a; eRF3 family member; eRF3c; ERF3-similar protein; ERFS; eukaryotic release factor 3; Hbs1; HBS1 like translational GTPase; HBS1L; Hbs1-like (S. cerevisiae); HBS1-like protein; HBS1-like translational GTPase; Hsp70 subfamily B suppressor 1-like protein; HSPC276; Kiaa1038
Mouse
Affinity chromatography
RUO
10767
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.