Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HBsAg (adr) (ACX36042, 227 amino acids) Full-length Recombinant Protein expressed in CHO cells

Catalog No. 89947128 Shop All Abnova Corporation Products
Click to view available options
Quantity:
50 μg

Used for SDS-PAGE

Sequence: MENTTSGFLGPLLVLQAGFFLLTRILTIPQSLDSWWTSLNFLGGAPTCPGQNSQSPTSNHSPTSCPPICPGYRWMCLRRFIIFLFILLLCLIFLLVLLDYQGMLPVCPLLPGTSTTSTGPCKTCTIPAQGTSMFPSCCCTKPSDGNCTCIPIPSSWAFARFLWEWASVRFSWLSLLVPFVQWFVGLSPTVWLSVIWMMWYWGPSLYNILSPFLPLLPIFFCLWVYI

Specifications

Accession Number ACX36042
For Use With (Application) SDS-PAGE
Formulation Liquid
Molecular Weight (g/mol) 24kDa
Name HBsAg (adr) Recombinant Protein
Preparation Method Mammalian cell (CHO) expression system
Purification Method Proprietary chromatography
Quantity 50 μg
Immunogen MENTTSGFLGPLLVLQAGFFLLTRILTIPQSLDSWWTSLNFLGGAPTCPGQNSQSPTSNHSPTSCPPICPGYRWMCLRRFIIFLFILLLCLIFLLVLLDYQGMLPVCPLLPGTSTTSTGPCKTCTIPAQGTSMFPSCCCTKPSDGNCTCIPIPSSWAFARFLWEWASVRFSWLSLLVPFVQWFVGLSPTVWLSVIWMMWYWGPSLYNILSPFLPLLPIFFCLWVYI
Storage Requirements Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name HBSAG
Species Hepatitis B
Recombinant Recombinant
Protein Tag None
Expression System Mammalian cell (CHO) expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.