Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBsAg Antibody (HB11), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1107391AF405
Description
HBsAg Monoclonal specifically detects HBsAg in Virus samples. It is validated for ELISA, Sandwich ELISA.Specifications
HBsAg | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
S | |
Recombinant full length protein (HBsAg of ayw subtype). | |
0.2 mL | |
Virology Bacteria and Parasites | |
944569 | |
Virus | |
Purified |
ELISA | |
HB11 | |
ELISA, Sandwich ELISA | |
HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reacts with the following HbsAg subtypes : ayw1, ayw2, ayw3, ayw4, ayr, adw2, adw4, adrq+, adrq-, ayw3 (Fer). | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction