Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBsAg Antibody (HB3), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500474B
Description
HBsAg Monoclonal antibody specifically detects HBsAg in Virus samples. It is validated for ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
HBsAg | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Virus | |
IgG1 |
ELISA, Immunocytochemistry | |
HB3 | |
ELISA, Immunocytochemistry/ Immunofluorescence | |
HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
Purified HBsAg from human plasma. | |
0.1 mL | |
Cell Biology, Virology Bacteria and Parasites | |
944569 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction